Ionomycin calcium salt 1 mg/ml in methanol, sterile filtered
Supplier: Thermo Scientific Chemicals
|
Danger
|
J67343-XF
10814-960EA
429.15
USD
10814-960
Ionomycin calcium salt 1 mg/ml in methanol, sterile filtered
Ionomycin calcium salt
Ionomycin calcium salt, 1 mg/mL in methanol, sterile-filtered.
Liquid
Caution: Research use only: Not intended for animal or human diagnostic or therapeutic use.
Formula:
C₄₁H₇₀CaO₉ Storage Temperature: Freezer |
MDL Number:
MFCD00151183 CAS Number: 56092-82-1 UN: 1230 ADR: "3",II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...