(+/-)13-Hydroxyoctadeca-9Z,11E-dienoic Acid ≥98% in ethanol

Danger

J67274-EXG
10814-954EA 249.41 USD
10814-954
(+/-)13-Hydroxyoctadeca-9Z,11E-dienoic Acid ≥98% in ethanol
(+/-)13-Hydroxyoctadeca-9Z,11E-dienoic Acid

  • Liquid

Formula: C₁₈H₃₂O₃
Storage Temperature: Freezer
MDL Number: MFCD00065832
CAS Number: 73804-64-5
UN: 1170
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR