Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Docosahexaenoyl 1,0% in ethanol , 98% stabilized (PAF C-16)

Supplier: Alfa Aesar
Danger
Synonyms: Ethanaminium, 2-[[[(2R)-3-(hexadecyloxy)-2-[[(4Z,7Z,10Z,13Z,16Z,19Z)-1-oxo-4,7,10,13,16,19-docosahexaen-1-yl]oxy]propoxy]hydroxyphosphinyl]oxy]-N,N,N-trimethyl-, inner salt(2R)-2-{[(4Z,7Z,10Z,13Z,16Z,19Z)-docosa-4,7,10,13,16,19-hexaenoyl]oxy}-3-(hexadecyloxy)propyl 2-(trimethylazaniumyl)ethyl phosphate1-Palmityl-2-docosahexaenoyl-sn-glycero-3-phosphocholine

Stabilization: Stabilised with BHT (Ionol) 0,1%

J67218-MCR J67218-LB0
10814-936EA 74.17 USD InStock
10814-936 10814-934
Docosahexaenoyl 1,0% in ethanol , 98% stabilized (PAF C-16)
Docosahexaenoyl


Formula: C₄₆H₈₂NO₇P
MW: 792.120 Da
Storage Temperature: Freezer
MDL Number: MFCD00216141
CAS Number: 132213-85-5
UN: 1170
ADR: 3,II
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty