Cinnamyl-3,4-dihydroxy-α-cyanocinnamate 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
J67167-MA
10816-070EA
178.55
USD
10816-070
Cinnamyl-3,4-dihydroxy-α-cyanocinnamate 98%
Cinnamyl-3,4-dihydroxy-alpha-cyanocinnamate
Formula:
C₁₉H₁₅NO₄ MW: 321.332 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00210839 CAS Number: 132465-11-3 UN: 3077 ADR: "9",III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...