(-)-Bicuculline methiodide 98%

Supplier: Thermo Scientific Chemicals
Danger

J67150-MF
10816-068EA 284.06 USD
10816-068
(-)-Bicuculline methiodide 98%
(-)-Bicuculline methiodide
Formula: C₂₁H₂₀INO₆
Storage Temperature: Ambient
MDL Number: MFCD00078966
CAS Number: 40709-69-1
UN: 1544
ADR: "6.1",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR