13(S)-Hydroxyoctadeca-9Z,11E-dienoic acid 90-100 ^mg/mL in ethanol , 98%
|
Danger
|
Synonyms:
S)-13-HODE
J67148-EXG
J67148-EXD
10816-064EA
310.95
USD
10816-064
10816-066
13(S)-Hydroxyoctadeca-9Z,11E-dienoic acid 90-100 ^mg/mL in ethanol , 98%
13(S)-Hydroxyoctadeca-9Z,11E-dienoic acid
Liquid
Formula:
C₁₈H₃₂O₃ MW: 296.45 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD00065833 CAS Number: 29623-28-7 EINECS: 200-578-6 UN: 1170 ADR: 3,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...