Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Tyrphostin AG 183 ≥98%

Supplier: Alfa Aesar
Warning
Synonyms: Tyrphostin A513-Amino-2,4-dicyano-5-(3',4',5'-trihydroxyphenyl)penta-2,4-dienonitrile

J67135-LB0 J67135-MB J67135-MA
10816-058EA 63.8 USD InStock
10816-058 10816-062 10816-060
Tyrphostin AG 183 ≥98%
Tyrphostin AG 183


Formula: C₁₃H₈N₄O₃
MW: 268.232 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00133903
CAS Number: 122520-90-5
UN: 3439
ADR: 6.1,III
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty