Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

α-Hydroxyfarnesylphosphonic acid 1 mg/500µl in ethanol , 96%

Supplier: Alfa Aesar
Danger

J67097-MCR
10816-054EA 243.24 USD InStock
10816-054
α-Hydroxyfarnesylphosphonic acid 1 mg/500µl in ethanol , 96%
α-Hydroxyfarnesylphosphonic acid
1 mg in 500µl ethanol


Formula: C15H27O4P
MW: 302.35 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00216148
CAS Number: 140633-12-1
UN: UN1170
ADR: 3,II
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty