N-Acetyl Cholecystokinin

Supplier: Thermo Scientific
J66989-EXD J66989-MCR
10816-038EA 121.27 USD
10816-038 10816-036
N-Acetyl Cholecystokinin
Proteins and Peptides
Molecular Formula: C38H50N8O13S3
Formula Weight: 923.04
Storage Temperature: -30°C to -10°C
Physical Form: Solid
Appearance: White to off-white
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR