Glu-Ile-Leu-Asp-Val

Supplier: Thermo Scientific
J66969-MCR
10815-944EA 50.78 USD
10815-944
Glu-Ile-Leu-Asp-Val
Proteins and Peptides
CAS No.: 150525-67-0
Molecular Formula: C26H45N5O10
Formula Weight: 587.66
Storage Temperature: -30°C to -10°C
Physical Form: Solid
Appearance: White to off-white
MDL No.: MFCD00144394
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR