C3 Peptide P16

Supplier: Thermo Scientific
J66952-EXD J66952-MCR
10815-928EA 110.06 USD
10815-928 10815-930
C3 Peptide P16
Proteins and Peptides
CAS No.: 99027-06-2
Molecular Formula: C86H131N25O27
Formula Weight: 1947.40
Storage Temperature: -30°C to -10°C
Physical Form: Solid
Appearance: White to off-white
MDL No.: MFCD00214596
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR