Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Protein Kinase C-epsilon Peptide Substrate

Supplier: Alfa Aesar
J66938-MCR J66938-EXD
10815-914EA 305.52 USD InStock
10815-914 10815-912
Protein Kinase C-epsilon Peptide Substrate
Proteins and Peptides
Molecular Formula: C83H15N39O21S
Formula Weight: 2067.45
Storage Temperature: -30°C to -10°C
Physical Form: Lyophilized powder
Appearance: White

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty