1,2,3,5-Tetra-O-benzoyl-2-C-methyl-β-D-ribofuranose 98%
Supplier: Thermo Scientific Chemicals
Synonyms:
(2S,3R,4R,5R)-5-((Benzoyloxy)methyl)-3-methyltetrahydrofuran-2,3,4-triyl tribenzoate
, (2S,3R,4R,5R)-2,4-bis(benzoyloxy)-5-[(benzoyloxy)methyl]-3-methyloxolan-3-yl benzoate
J66930-03
J66930-06
10815-904EA
113.63
USD
10815-904
10815-902
1,2,3,5-Tetra-O-benzoyl-2-C-methyl-β-D-ribofuranose 98%
1,2,3,5-Tetra-O-benzoyl-2-C-methyl-beta-D-ribofuranose
Formula:
C₃₄H₂₈O₉ MW: 580.589 g/mol Melting Pt: 147…163 °C Storage Temperature: Freezer |
MDL Number:
MFCD07369658 CAS Number: 15397-15-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...