N-acetyl-pepstatin

Supplier: Thermo Scientific
J66917-MF J66917-MB
10814-918EA 263.49 USD
10814-918 10814-920
N-acetyl-pepstatin
CAS No.: 28575-34-0
Molecular Formula: C31H57N5O9
Formula Weight: 643.82
Storage Temperature: -30°C to -10°C
Physical Form: Solid
Appearance: White to off-white
MDL No.: MFCD00214071
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR