1,6-Anhydro-β-D-cellobiose 95%

Supplier: Thermo Scientific Chemicals
Warning

J66914-ME J66914-03 J66914-06
10814-912EA 91.82 USD
10814-912 10814-910 10814-908
1,6-Anhydro-β-D-cellobiose 95%
1,6-Anhydro-beta-D-cellobiose
Formula: C₁₂H₂₀O₁₀
MW: 324.282 g/mol
Storage Temperature: Ambient
MDL Number: MFCD13182343
CAS Number: 35405-71-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR