Caspase-3-substrate III
Supplier: Thermo Scientific Chemicals
J66881-EXD
J66881-MCR
10814-888EA
301.3
USD
10814-888
10814-886
Caspase-3-substrate III
Caspase-3-substrate III
Formula:
C₅₀H₆₂N₁₀O₂₂ MW: 1155.094 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD00798785 CAS Number: 189696-20-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...