Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Furoyl-Leu-Ile-Gly-Arg-Leu-Orn-NH_2

Supplier: Alfa Aesar
J66850-LB0 J66850-MCR
10815-898EA 297.44 USD InStock
10815-898 10814-860
2-Furoyl-Leu-Ile-Gly-Arg-Leu-Orn-NH_2
Proteins and Peptides
CAS No.: 729589-58-6
Molecular Formula: C36H63N11O8
Formula Weight: 777.97
Storage Temperature: -30°C to -10°C
Physical Form: Lyophilized powder
Appearance: White
MDL No.: MFCD08460329

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty