Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Cecropin A (1-8)-Melittin (1-18) amide

Supplier: Alfa Aesar
J66814-MCR J66814-EXD
10815-862EA 300.86 USD InStock
10815-862 10815-860
Cecropin A (1-8)-Melittin (1-18) amide
Proteins and Peptides
Molecular Formula: C136H233N33O29
Formula Weight: 2794.50
Storage Temperature: -30°C to -10°C
Physical Form: Lyophilized powder
Appearance: White to off-white

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty