Gly-D-Arg-Gly-Asp-Ser-Pro-Ala-Ser-Ser-Lys

Supplier: Thermo Scientific
J66799-MCR
10815-838EA 99.23 USD
10815-838
Gly-D-Arg-Gly-Asp-Ser-Pro-Ala-Ser-Ser-Lys
Proteins and Peptides
Molecular Formula: C37H64N14O16
Formula Weight: 960.99
Storage Temperature: -30°C to -10°C
Physical Form: Solid
Appearance: White to off-white
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR