N-Acetyl-Leu-Glu-His-Asp-Al

Supplier: Thermo Scientific
J66787-MCR J66787-LB0
10814-848EA 242.62 USD
10814-848 10814-850
N-Acetyl-Leu-Glu-His-Asp-Al
Proteins and Peptides
Molecular Formula: C23H34N6O9
Formula Weight: 538.55
Storage Temperature: -30°C to -10°C
Physical Form: Lyophilized powder
Appearance: White to off-white
MDL No.: MFCD01862665

  • Light Sensitive
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR