Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Caspase-3 Substrate V, fluorogenic

Supplier: Alfa Aesar
J66767-EXD J66767-MCR
10814-844EA 121.16 USD InStock
10814-844 10814-846
Caspase-3 Substrate V, fluorogenic
Proteins and Peptides
Molecular Formula: C60H75N14O21S
Formula Weight: 1360.38
Storage Temperature: -30°C to -10°C
Physical Form: Lyophilized powder
Appearance: White to off-white

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty