1,2-O-Cyclohexylidene-α-D-glucofuranose 97%
J66761-03
J66761-06
10814-842EA
185.02
USD
10814-842
10814-840
1,2-O-Cyclohexylidene-α-D-glucofuranose 97%
1,2-O-Cyclohexylidene-alpha-D-glucofuranose
Formula:
C₁₂H₂₀O₆ Melting Pt: 163…168 °C Storage Temperature: Ambient |
MDL Number:
MFCD00142481 CAS Number: 16832-21-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...