Glu-Ile-Leu-Glu-Val-Pro-Ser-Thr

Supplier: Thermo Scientific
J66722-LB0
10814-794EA 246.38 USD
10814-794
Glu-Ile-Leu-Glu-Val-Pro-Ser-Thr
Proteins and Peptides
Molecular Formula: C39H66N8O15
Formula Weight: 886.98
Storage Temperature: -30°C to -10°C
Physical Form: Solid
Appearance: White to off-white
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR