Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

N-Acetyl-Val-Ala-Asp-al

Supplier: Alfa Aesar
J66696-MCR J66696-LB0
10814-780EA 56.23 USD InStock
10814-780 10814-778
N-Acetyl-Val-Ala-Asp-al
Proteins and Peptides
CAS No.: 147837-52-3
Molecular Formula: C14H23N3O6
Formula Weight: 329.35
Storage Temperature: -30°C to -10°C
Physical Form: Lyophilized powder
Appearance: White to off-white
MDL No.: MFCD00672421

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty