4-Methylphenyl-β-D-thiogalactopyranoside 98%
Supplier: Thermo Scientific Chemicals
Synonyms:
4-Methylphenyl 1-thio-beta-D-galactopyranoside
J66686-06
J66686-03
10814-764EA
251.89
USD
10814-764
10814-762
4-Methylphenyl-β-D-thiogalactopyranoside 98%
4-Methylphenyl-beta-D-thiogalactopyranoside
Formula:
C₁₃H₁₈O₅S MW: 286.342 g/mol Melting Pt: 137…139 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD04973631 CAS Number: 28244-98-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...