4-Methylumbelliferyl-β-D-glucopyranoside 99%

Supplier: Thermo Scientific Chemicals

Synonyms: 4-Methylumbelliferyl beta-D-GlucopyranosideMUGlucosid4-MU-β-D-Glc

J66630-MD J66630-03 J66630-06
10815-812EA 484.82 USD
10815-812 10815-814 10815-810
4-Methylumbelliferyl-β-D-glucopyranoside 99%
4-Methylumbelliferyl-beta-D-glucopyranoside
Formula: C₁₆H₁₈O₈
MW: 338.312 g/mol
Melting Pt: 210…212 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00063694
CAS Number: 18997-57-4
EINECS: 242-736-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR