Human Neuropeptide Y,Biotin

Supplier: Thermo Scientific
J66629-MCR
10815-808EA 322.59 USD
10815-808
Human Neuropeptide Y,Biotin
Proteins and Peptides
Molecular Formula: C199H300N57O60S2
Formula Weight: 4515.04
Storage Temperature: -30°C to -10°C
Physical Form: Solid
Appearance: White to off-white
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR