4,6-O-Ethylidene-ɑ-D-glucopyranose 90%

Supplier: Thermo Scientific Chemicals

Synonyms: 4,6-O-Ethylidene-alpha-D-glucose

J66583-06 J66583-03
10814-722EA 225.19 USD
10814-722 10814-720
4,6-O-Ethylidene-ɑ-D-glucopyranose 90%
4,6-O-Ethylidene-alpha-D-glucopyranose
Formula: C₈H₁₄O₆
MW: 206.19 g/mol
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00006820
CAS Number: 13224-99-2
EINECS: 236-198-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR