Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Gly-Pen-Gly-Arg-Gly-Asp-Ser-Pro-Cys-Ala (Disulfide bridge: Pen2-Cys9)

Supplier: Alfa Aesar
J66560-MCR
10814-696EA 116.59 USD InStock
10814-696
Gly-Pen-Gly-Arg-Gly-Asp-Ser-Pro-Cys-Ala (Disulfide bridge: Pen2-Cys9)
Proteins and Peptides
CAS No.: 111844-17-8
Molecular Formula: C35H57N13O14S2
Formula Weight: 948.03
Storage Temperature: -30°C to -10°C
Physical Form: Solid
Appearance: White to off-white
MDL No.: MFCD00133377

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty