N-Acetyl-Adhesin (1025-1044)

Supplier: Thermo Scientific
J66492-EXD J66492-MCR
10815-592EA 164.16 USD
10815-592 10815-594
N-Acetyl-Adhesin (1025-1044)
Proteins and Peptides
Molecular Formula: C97H160N26O32
Formula Weight: 2202.46
Storage Temperature: -30°C to -10°C
Physical Form: Lyophilized powder
Appearance: White to off-white
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR