D-Fructose-16-diphosphate dicalcium salt 95%

Supplier: Thermo Scientific Chemicals

J66477-30 J66477-22 J66477-14
10815-578EA 331.75 USD
10815-578 10814-664 10814-662
D-Fructose-16-diphosphate dicalcium salt 95%
D-Fructose-16-diphosphate dicalcium salt
Formula: C₆H₁₀Ca₂O₁₂P₂
Storage Temperature: Ambient
MDL Number: MFCD00135875
CAS Number: 6055-82-9
EINECS: 227-979-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR