[D-Arg2] Dermorphin (1-4)

Supplier: Thermo Scientific
J66448-MCR J66448-LB0
10814-656EA 50.78 USD
10814-656 10814-654
[D-Arg2] Dermorphin (1-4)
Proteins and Peptides
CAS No.: 100304-60-7
Molecular Formula: C26H36N8O5
Formula Weight: 540.62
Storage Temperature: -30°C to -10°C
Physical Form: Solid
Appearance: White to off-white
MDL No.: MFCD00076697
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR