Glu-Glu-Lys-Leu-Ile-Val-Val-Ala-Phe

Supplier: Thermo Scientific
J66445-MCR
10814-652EA 70.68 USD
10814-652
Glu-Glu-Lys-Leu-Ile-Val-Val-Ala-Phe
Proteins and Peptides
CAS No.: 188818-21-5
Molecular Formula: C50H82N10O14
Formula Weight: 1047.30
Storage Temperature: -30°C to -10°C
Physical Form: Solid
Appearance: White to off-white
MDL No.: MFCD00798761
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR