Decyl-β-D-glucopyranoside

Supplier: Thermo Scientific Chemicals

Synonyms: N-Decyl-b-D-glucopyranosideDecyl GlucopyranosideDecyl beta-D-glucopyranoside

J66423-03 J66423-06
10814-632EA 539.2 USD
10814-632 10814-630
Decyl-β-D-glucopyranoside
Decyl β-D-glucopyranoside

Non-ionic detergent used for solubilizing membrane proteins and in the development of sugar-based surfactant solutions.

Formula: C₁₆H₃₂O₆
MW: 320.43 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00063297
CAS Number: 58846-77-8
EINECS: 261-469-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR