4-Methoxyphenyl-β-D-glucopyranoside 98%
Supplier: Thermo Scientific Chemicals
Synonyms:
(2R,3S,4S,5R,6S)-2-(Hydroxymethyl)-6-(4-methoxyphenoxy)tetrahydro-2H-pyran-3,4,5-triol
J66422-03
J66422-06
10814-628EA
620.44
USD
10814-628
10814-626
4-Methoxyphenyl-β-D-glucopyranoside 98%
4-Methoxyphenyl-beta-D-glucopyranoside
Formula:
C₁₃H₁₈O₇ MW: 286.28 g/mol Melting Pt: 175…179 °C Storage Temperature: Ambient |
MDL Number:
MFCD06797143 CAS Number: 6032-32-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...