1,2:5,6-Di-O-isopropylidene-α-D-allofuranose 98%

Supplier: Thermo Scientific Chemicals

Synonyms: 1,2:5,6-Di-o-isopropylidene-alpha-D-allofuranose

J66388-03 J66388-06
10814-608EA 72.94 USD
10814-608 10814-610
1,2:5,6-Di-O-isopropylidene-α-D-allofuranose 98%
1,2:5,6-Di-O-isopropylidene-alpha-D-allofuranose
Formula: C12H20O6
MW: 260.28 g/mol
Melting Pt: 73…76 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00135634
CAS Number: 2595-05-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR