Phenyl-2,3,4,6-tetra-O-acetyl-1-thio-β-D-glucopyranoside 98%
Supplier: Thermo Scientific Chemicals
Synonyms:
(2R,3R,4S,5R,6S)-2-(Acetoxymethyl)-6-(phenylthio)tetrahydro-2H-pyran-3,4,5-triyl triacetate
, 1-Phenyl-1-thio-β-D-glucopyranoside tetraacetate
, Phenyl 2,3,4,6-tetra-o-acetyl-1-thio-β-D-glucopyranoside
J66355-03
J66355-06
10814-576EA
257.89
USD
10814-576
10814-574
Phenyl-2,3,4,6-tetra-O-acetyl-1-thio-β-D-glucopyranoside 98%
Phenyl-2,3,4,6-tetra-O-acetyl-1-thio-beta-D-glucopyranoside
Formula:
C₂₀H₂₄O₉S MW: 440.46 g/mol Melting Pt: 116…119 °C Storage Temperature: Ambient |
MDL Number:
MFCD00135159 CAS Number: 23661-28-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...