R15K, Hiv-1 Inhibitory Peptide

Supplier: Thermo Scientific
J66327-EXD J66327-MCR
10815-562EA 50.78 USD
10815-562 10815-564
R15K, Hiv-1 Inhibitory Peptide
Proteins and Peptides
CAS No.: 114991-28-5
Molecular Formula: C73H126N26O18
Formula Weight: 1655.97
Storage Temperature: -30°C to -10°C
Physical Form: Solid
Appearance: White to off-white
MDL No.: MFCD00235749
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR