DPDPE

Supplier: Alfa Aesar

J66293-LB0 J66293-MCR
10815-490EA 228.85 USD
10815-490 10815-492
DPDPE
DPDPE
Formula: C₃₀H₃₉N₅O₇S₂
MW: 645.79 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00076430
CAS Number: 88373-73-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR