Human N-Acetyl-[D-Trp16] Endothelin-1 (16-21)

Supplier: Thermo Scientific
J66214-MB J66214-MA
10814-518EA 309.75 USD
10814-518 10814-520
Human N-Acetyl-[D-Trp16] Endothelin-1 (16-21)
Proteins and Peptides
CAS No.: 143037-33-6
Molecular Formula: C46H62N8O10
Formula Weight: 887.03
Storage Temperature: -30°C to -10°C
Physical Form: Solid
Appearance: White to off-white
MDL No.: MFCD00236979
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR