Human [Glu1]-Gastrin I,Biotin

Supplier: Thermo Scientific
J66203-MCR J66203-EXD
10814-516EA 199.26 USD
10814-516 10814-514
Human [Glu1]-Gastrin I,Biotin
Molecular Formula: C107H140N22O34S2
Formula Weight: 2342.60
Storage Temperature: -30°C to -10°C
Physical Form: Solid
Appearance: White to off-white
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR