Sucrose diacetate hexaisobutyrate

Supplier: Thermo Scientific Chemicals

Synonyms: SAIB

J66195-A1 J66195-36 J66195-30
10814-508EA 261.54 USD
10814-508 10814-506 10814-504
Sucrose diacetate hexaisobutyrate
Sucrose diacetate hexaisobutyrate
Formula: C₄₀H₆₂O₁₉
MW: 846.91
Boiling Pt: 288 °C
Melting Pt: –7 °C
Density: 1,146
Storage Temperature: Ambient
MDL Number: MFCD00041859
CAS Number: 27216-37-1
EINECS: 204-771-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR