β-D-Allose 97%
Synonyms:
β-D-Allopyranose
J66172-MD
J66172-03
10814-476EA
81.5
USD
10814-476
10814-474
β-D-Allose 97%
β-D-Allose
Formula:
C₆H₁₂O₆ MW: 180.16 g/mol Melting Pt: 148…150 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00063268 CAS Number: 7283-09-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...