Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

N-Acetyl-Asp-Tyr(2-malonyl)-Val-Pro-Met-Leu-NH_2

Supplier: Alfa Aesar
J66171-MCR J66171-EXD
10814-472EA 121.16 USD InStock
10814-472 10814-470
N-Acetyl-Asp-Tyr(2-malonyl)-Val-Pro-Met-Leu-NH_2
Proteins and Peptides
CAS No.: 168135-79-3
Molecular Formula: C39H57N7O14S
Formula Weight: 879.98
Storage Temperature: -30°C to -10°C
Physical Form: Solid
Appearance: White to off-white
MDL No.: MFCD00798790

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty