[Des-Tyr1] gamma-Endorphin

J66144-MCR
10814-452EA 80.06 USD
10814-452
[Des-Tyr1] gamma-Endorphin
[Des-Tyr1] gamma-Endorphin
Formula: C₇₄H₁₂₂N₁₈O₂₅S
MW: 1695.951 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00076397
CAS Number: 67810-56-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR