2,3,4,6-Tetra-O-acetyl-α-D-mannopyranosyl fluoride 98%

Synonyms: (2R,3R,4S,5S,6R)-2-(Acetoxymethyl)-6-fluorotetrahydro-2H-pyran-3,4,5-triyl triacetate

J66137-03 J66137-06
10814-448EA 391.43 USD
10814-448 10814-446
2,3,4,6-Tetra-O-acetyl-α-D-mannopyranosyl fluoride 98%
2,3,4,6-Tetra-O-acetyl-alpha-D-mannopyranosyl fluoride
Formula: C₁₄H₁₉FO₉
MW: 350.295 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00792706
CAS Number: 2823-44-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR