Phenyl-2,3,4,6-tetra-O-acetyl-1-thio-β-D-galactopyranoside 98%
Synonyms:
(2R,3S,4S,5R,6S)-2-(Acetoxymethyl)-6-(phenylthio)tetrahydro-2H-pyran-3,4,5-triyl triacetate
J66122-03
10815-464EA
36.51
USD
10815-464
Phenyl-2,3,4,6-tetra-O-acetyl-1-thio-β-D-galactopyranoside 98%
Phenyl-2,3,4,6-tetra-O-acetyl-1-thio-beta-D-galactopyranoside
Formula:
C₂₀H₂₄O₉S MW: 440.46 g/mol Melting Pt: 68…72 °C Storage Temperature: Freezer |
MDL Number:
MFCD06657849 CAS Number: 24404-53-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...