Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

[D-Lys3] Growth Hormone Releasing Hexapeptide

Supplier: Alfa Aesar
J66103-LB0 J66103-MB
10815-440EA 119.89 USD InStock
10815-440 10815-442
[D-Lys3] Growth Hormone Releasing Hexapeptide
Proteins and Peptides
CAS No.: 136054-22-3
Molecular Formula: C49H63N13O6
Formula Weight: 930.12
Storage Temperature: -30°C to -10°C
Physical Form: Lyophilized powder
Appearance: White to off-white
Solubility: Soluble in water.
MDL No.: MFCD00214665

Antagonist for GHRP-6

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty