Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

N-Acetyl-Val-Asp-Val-Ala-Asp-7-amino-4-methylcoumarin

Supplier: Alfa Aesar
J66089-LB0 J66089-MB
10814-422EA 219.21 USD InStock
10814-422 10814-424
N-Acetyl-Val-Asp-Val-Ala-Asp-7-amino-4-methylcoumarin
Proteins and Peptides
Molecular Formula: C33H44N6O12
Formula Weight: 716.73
Storage Temperature: -30°C to -10°C
Physical Form: Lyophilized powder
Appearance: White to off-white

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty