Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

N-Acetyl-Ser-Gln-Asn-Tyr-Pro-Val-Val-NH_2

Supplier: Alfa Aesar
J66080-MCR J66080-LB0
10814-416EA 44.54 USD InStock
10814-416 10814-414
N-Acetyl-Ser-Gln-Asn-Tyr-Pro-Val-Val-NH_2
Proteins and Peptides
CAS No.: 121822-32-0
Molecular Formula: C38H58N10O12
Formula Weight: 846.94
Storage Temperature: -30°C to -10°C
Physical Form: Solid
Appearance: White to off-white
MDL No.: MFCD00133021

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty